CAS RN 5176-27-2
IUPAC Name:
tert-butyl 1H-pyrrole-1-carboxylate
Synonyms:
tert-butyl pyrrole-1-carboxylate
Molecular Weight (g/mol):
167.21
Molecular Formula:
C9H13NO2
InChi Key:
IZPYBIJFRFWRPR-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)N1C=CC=C1
1
–
1
of
1
results
1
Promotions Available
N-BOC-Pyrrole, 98%
N-BOC-Pyrrole, 98%
CAS: 5176-27-2 Molecular Formula: C9H13NO2 Molecular Weight (g/mol): 167.21 InChI Key: IZPYBIJFRFWRPR-UHFFFAOYSA-N Synonym: tert-butyl 1h-pyrrole-1-carboxylate,n-boc-pyrrole,n-t-boc-pyrrole,tert-butyl 1-pyrrolecarboxylate,t-butyl 1h-pyrrole-1-carboxylate,1h-pyrrole-1-carboxylic acid, 1,1-dimethylethyl ester,n-tert-butoxycarbonyl-1h-pyrrole,1-boc pyrrole,1-boc-pyrrole,acmc-209kvp PubChem CID: 643494 IUPAC Name: tert-butyl pyrrole-1-carboxylate SMILES: CC(C)(C)OC(=O)N1C=CC=C1