missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-p-phenylenediamine, 97%

Catalog No. AC398330010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398330010 1 g Glass bottle
AC398330050 5 g Glass bottle
AC398330250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC398330010 Supplier Thermo Scientific Chemicals Supplier No. 398330010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 71026-66-9 | C11H16N2O2 | 208.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71026-66-9
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.26
MDL Number MFCD00043022
InChI Key WIVYTYZCVWHWSH-UHFFFAOYSA-N
PubChem CID 688611
IUPAC Name tert-butyl N-(4-aminophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 113°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym tert-butyl 4-aminophenyl carbamate, 4-tert-butoxycarbonylamino aniline, tert-butyl 4-aminophenylcarbamate, tert-butyl n-4-aminophenyl carbamate, n-boc-p-phenylenediamine, n-boc-1,4-phenylene diamine, n-boc-1,4-phenylenediamine, 4-amino-phenyl-carbamic acid tert-butyl ester, tert-butyl4-aminophenylcarbamate, 4-t-butyloxycarbonylamino aniline
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material N-BOC-p-phenylenediamine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 275-132-7

RUO – Research Use Only