missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-O-tert-butyl-L-serine, 98%
CAS: 13734-38-8 | C12H23NO5 | 261.318 g/mol
$106.44 - $370.71
Chemical Identifiers
| CAS | 13734-38-8 |
|---|---|
| Molecular Formula | C12H23NO5 |
| Molecular Weight (g/mol) | 261.318 |
| MDL Number | MFCD00079666 |
| InChI Key | BPYLRGKEIUPMRJ-QMMMGPOBSA-N |
| Synonym | boc-ser tbu-oh, boc-o-tert-butyl-l-serine, boc-d-ser tbu-oh, l-serine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-tert-butoxy-2-tert-butoxycarbonyl amino propanoic acid, boc-ser but-oh, boc-l-ser tbu, boc-hser bzl-oh, pubchem12157, boc-o-t-butyl-l-serine |
| PubChem CID | 7017896 |
| IUPAC Name | (2S)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| SMILES | CC(C)(C)OCC(C(=O)O)NC(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6214703
|
Thermo Scientific Chemicals
H6214703 |
1 g |
Each for $106.44
|
|
|||||
|
AAH6214706
|
Thermo Scientific Chemicals
H6214706 |
5 g |
Each for $370.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13734-38-8 | |
| 261.318 | |
| BPYLRGKEIUPMRJ-QMMMGPOBSA-N | |
| 7017896 | |
| CC(C)(C)OCC(C(=O)O)NC(=O)OC(C)(C)C |
| C12H23NO5 | |
| MFCD00079666 | |
| boc-ser tbu-oh, boc-o-tert-butyl-l-serine, boc-d-ser tbu-oh, l-serine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-tert-butoxy-2-tert-butoxycarbonyl amino propanoic acid, boc-ser but-oh, boc-l-ser tbu, boc-hser bzl-oh, pubchem12157, boc-o-t-butyl-l-serine | |
| (2S)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
| 13734-38-8 | |
| MFCD00079666 | |
| BPYLRGKEIUPMRJ-QMMMGPOBSA-N | |
| (2S)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 261.318 | |
| 261.31 | |
| N-Boc-O-tert-butyl-L-serine |
| C12H23NO5 | |
| boc-ser tbu-oh, boc-o-tert-butyl-l-serine, boc-d-ser tbu-oh, l-serine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-tert-butoxy-2-tert-butoxycarbonyl amino propanoic acid, boc-ser but-oh, boc-l-ser tbu, boc-hser bzl-oh, pubchem12157, boc-o-t-butyl-l-serine | |
| CC(C)(C)OCC(C(=O)O)NC(=O)OC(C)(C)C | |
| 1 g | |
| 7017896 | |
| 98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only