accessibility menu, dialog, popup

UserName

CAS RN 13734-38-8

H3CCH3H3CONHOOCH3CH3H3COHO

CAS RN 13734-38-8

IUPAC Name: 3-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 3-(tert-butoxy)-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 261.32
Molecular Formula: C12H23NO5
InChi Key: BPYLRGKEIUPMRJ-UHFFFAOYNA-N
SMILES: CC(C)(C)OCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)