missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-tert-butyl-L-serine, 98%

Catalog No. AAH6214703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6214703 1 g
AAH6214706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6214703 Supplier Thermo Scientific Chemicals Supplier No. H6214703

CAS: 13734-38-8 | C12H23NO5 | 261.318 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13734-38-8
Molecular Formula C12H23NO5
Molecular Weight (g/mol) 261.318
MDL Number MFCD00079666
InChI Key BPYLRGKEIUPMRJ-QMMMGPOBSA-N
Synonym boc-ser tbu-oh, boc-o-tert-butyl-l-serine, boc-d-ser tbu-oh, l-serine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-tert-butoxy-2-tert-butoxycarbonyl amino propanoic acid, boc-ser but-oh, boc-l-ser tbu, boc-hser bzl-oh, pubchem12157, boc-o-t-butyl-l-serine
PubChem CID 7017896
IUPAC Name (2S)-3-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Quantity 1 g
Formula Weight 261.31
Percent Purity 98%
Chemical Name or Material N-Boc-O-tert-butyl-L-serine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only