missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-O-ethyl-L-serine, 97%
CAS: 104839-00-1 | C10H19NO5 | 233.264 g/mol
$277.15 - $912.66
Chemical Identifiers
| CAS | 104839-00-1 |
|---|---|
| Molecular Formula | C10H19NO5 |
| Molecular Weight (g/mol) | 233.264 |
| MDL Number | MFCD06797558 |
| InChI Key | KNBDVSSESAKBJM-ZETCQYMHSA-N |
| Synonym | boc-s-2-amino-3-ethoxypropionic acid, s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, 2s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, boco-etser, serine, n-1,1-dimethylethoxy carbonyl-o-ethyl, n-tert-butoxycarbonyl-o-ethylserine, n-boc-2-amino-3-ethoxy-propionic acid, s-n-boc-2-amino-3-ethoxypropionic acid, s-2-tertbutoxycarbonylamino-3-ethoxypropionic acid, n-1,1-dimethylethyl oxy carbonyl-o-ethylserine |
| PubChem CID | 15058451 |
| IUPAC Name | (2S)-3-ethoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| SMILES | CCOCC(C(=O)O)NC(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52781MD
|
Thermo Scientific Chemicals
H52781MD |
250 mg |
Each for $277.15
|
|
|||||
|
AAH5278103
|
Thermo Scientific Chemicals
H5278103 |
1 g |
Each for $912.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 104839-00-1 | |
| 233.264 | |
| KNBDVSSESAKBJM-ZETCQYMHSA-N | |
| 15058451 | |
| CCOCC(C(=O)O)NC(=O)OC(C)(C)C |
| C10H19NO5 | |
| MFCD06797558 | |
| boc-s-2-amino-3-ethoxypropionic acid, s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, 2s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, boco-etser, serine, n-1,1-dimethylethoxy carbonyl-o-ethyl, n-tert-butoxycarbonyl-o-ethylserine, n-boc-2-amino-3-ethoxy-propionic acid, s-n-boc-2-amino-3-ethoxypropionic acid, s-2-tertbutoxycarbonylamino-3-ethoxypropionic acid, n-1,1-dimethylethyl oxy carbonyl-o-ethylserine | |
| (2S)-3-ethoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
| 104839-00-1 | |
| MFCD06797558 | |
| KNBDVSSESAKBJM-ZETCQYMHSA-N | |
| (2S)-3-ethoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 233.264 | |
| 233.26 | |
| N-Boc-O-ethyl-L-serine |
| C10H19NO5 | |
| boc-s-2-amino-3-ethoxypropionic acid, s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, 2s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, boco-etser, serine, n-1,1-dimethylethoxy carbonyl-o-ethyl, n-tert-butoxycarbonyl-o-ethylserine, n-boc-2-amino-3-ethoxy-propionic acid, s-n-boc-2-amino-3-ethoxypropionic acid, s-2-tertbutoxycarbonylamino-3-ethoxypropionic acid, n-1,1-dimethylethyl oxy carbonyl-o-ethylserine | |
| CCOCC(C(=O)O)NC(=O)OC(C)(C)C | |
| 250 mg | |
| 15058451 | |
| 97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only