accessibility menu, dialog, popup

UserName

H3CO(S)HNOOCH3CH3CH3HOO

CAS RN 104839-00-1

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-ethoxypropanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-ethoxypropanoic acid
Molecular Weight (g/mol): 233.26
Molecular Formula: C10H19NO5
InChi Key: KNBDVSSESAKBJM-ZETCQYMHSA-N
SMILES: CCOC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)