missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-ethyl-L-serine, 97%

Catalog No. AAH5278103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5278103 1 g
AAH52781MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5278103 Supplier Thermo Scientific Chemicals Supplier No. H5278103
Only null left

CAS: 104839-00-1 | C10H19NO5 | 233.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104839-00-1
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.264
MDL Number MFCD06797558
InChI Key KNBDVSSESAKBJM-ZETCQYMHSA-N
Synonym boc-s-2-amino-3-ethoxypropionic acid, s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, 2s-2-tert-butoxycarbonyl amino-3-ethoxypropanoic acid, boco-etser, serine, n-1,1-dimethylethoxy carbonyl-o-ethyl, n-tert-butoxycarbonyl-o-ethylserine, n-boc-2-amino-3-ethoxy-propionic acid, s-n-boc-2-amino-3-ethoxypropionic acid, s-2-tertbutoxycarbonylamino-3-ethoxypropionic acid, n-1,1-dimethylethyl oxy carbonyl-o-ethylserine
PubChem CID 15058451
IUPAC Name (2S)-3-ethoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CCOCC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Quantity 1 g
Formula Weight 233.26
Percent Purity 97%
Chemical Name or Material N-Boc-O-ethyl-L-serine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only