missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-benzyl-L-threonine, 99%

Catalog No. AAB2167703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2167703 1 g
AAB2167706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2167703 Supplier Thermo Scientific Chemicals Supplier No. B2167703

CAS: 15260-10-3 | C16H23NO5 | 309.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15260-10-3
Molecular Formula C16H23NO5
Molecular Weight (g/mol) 309.36
MDL Number MFCD00066062
InChI Key CTXPLTPDOISPTE-YPMHNXCESA-N
Synonym boc-thr bzl-oh, boc-o-benzyl-l-threonine, n-tert-butoxycarbonyl-o-benzyl-l-threonine, n-boc-o-benzyl-l-threonine, boc-thr bzl, o-benzyl-n-tert-butoxycarbonyl-l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-threonine, l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2s,3r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxybutanoic acid
PubChem CID 1549483
IUPAC Name (2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
SMILES C[C@@H](OCC1=CC=CC=C1)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 115°C to 116°C
Quantity 1 g
Formula Weight 309.36
Percent Purity 99%
Chemical Name or Material N-Boc-O-benzyl-L-threonine
EINECSNumber 239-304-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only