missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-O-Benzyl-D-serine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 47173-80-8
Molecular Formula C15H20NO5
Molecular Weight (g/mol) 294.33
MDL Number MFCD00038248
InChI Key DMBKPDOAQVGTST-GFCCVEGCSA-M
Synonym n-boc-o-benzyl-d-serine, boc-d-ser bzl-oh, boc-o-benzyl-d-serine, o-benzyl-n-tert-butoxycarbonyl-d-serine, d-serine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2r-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid, pubchem12444, pubchem14944, boc-d-ser obn-oh
PubChem CID 2733693
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC334670050
SDS View Documents
Thermo Scientific Chemicals
334670050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC334670250
SDS View Documents
Thermo Scientific Chemicals
334670250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

47173-80-8
294.33
DMBKPDOAQVGTST-GFCCVEGCSA-M
2733693
CC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O
C15H20NO5
MFCD00038248
n-boc-o-benzyl-d-serine, boc-d-ser bzl-oh, boc-o-benzyl-d-serine, o-benzyl-n-tert-butoxycarbonyl-d-serine, d-serine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2r-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid, pubchem12444, pubchem14944, boc-d-ser obn-oh
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid

Specifications

47173-80-8
100.0
97.5% min. (HPLC)
C15H20NO5
5g
-19
DMBKPDOAQVGTST-GFCCVEGCSA-M
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid
2733693
98%
97.5
Authentic
Glass bottle
MFCD00038248
−19.00 (20.00°C c=2 80% alcohol)
n-boc-o-benzyl-d-serine, boc-d-ser bzl-oh, boc-o-benzyl-d-serine, o-benzyl-n-tert-butoxycarbonyl-d-serine, d-serine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2r-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid, pubchem12444, pubchem14944, boc-d-ser obn-oh
CC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O
294.33
295.33
N-BOC-O-Benzyl-D-serine, 98%
Videos
SDS
Documents

Documents

RUO – Research Use Only