Learn More
N-Boc-N-methylethanolamine, 95%
CAS: 57561-39-4 | C8H17NO3 | 175.228 g/mol
Supplier: Thermo Scientific Chemicals H6666006
| Quantity | 5 g |
|---|
Description
N-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.
Solubility
Soluble in diochloromethane. Slightly soluble in water.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 57561-39-4 | |
| 175.228 | |
| RFDSJHHLGFFVHD-UHFFFAOYSA-N | |
| 545700 | |
| CC(C)(C)OC(=O)N(C)CCO |
| C8H17NO3 | |
| MFCD03425857 | |
| n-boc-n-methyl-aminoethanol, tert-butyl 2-hydroxyethyl methyl carbamate, boc,me-glycinol, tert-butyl n-2-hydroxyethyl-n-methylcarbamate, 2-n-methyl-n-boc-amino ethanol, 2-n-boc-n-methylamino ethanol, n-boc-n-methylethanolamine, 2-hydroxyethyl methylcarbamic acid 1,1-dimethylethyl ester, carbamic acid, n-2-hydroxyethyl-n-methyl-, 1,1-dimethylethyl ester | |
| tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate |
Specifications
| N-Boc-N-methylethanolamine | |
| Colorless to Yellow | |
| MFCD03425857 | |
| n-boc-n-methyl-aminoethanol, tert-butyl 2-hydroxyethyl methyl carbamate, boc,me-glycinol, tert-butyl n-2-hydroxyethyl-n-methylcarbamate, 2-n-methyl-n-boc-amino ethanol, 2-n-boc-n-methylamino ethanol, n-boc-n-methylethanolamine, 2-hydroxyethyl methylcarbamic acid 1,1-dimethylethyl ester, carbamic acid, n-2-hydroxyethyl-n-methyl-, 1,1-dimethylethyl ester | |
| RFDSJHHLGFFVHD-UHFFFAOYSA-N | |
| tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate | |
| 175.228 | |
| 175.23 | |
| Liquid |
| 57561-39-4 | |
| C8H17NO3 | |
| 3537964 | |
| Soluble in diochloromethane. Slightly soluble in water. | |
| CC(C)(C)OC(=O)N(C)CCO | |
| 5 g | |
| 545700 | |
| 95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C