missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methylethanolamine, 95%

Catalog No. AAH6666003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6666003 1 g
AAH6666006 5 g
AAH6666014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6666003 Supplier Thermo Scientific Chemicals Supplier No. H6666003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57561-39-4 | C8H17NO3 | 175.228 g/mol

N-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-N-methylethanolamine is an substituent in the synthesis of anticancer agent, an orally bioavailable small molecule inhibitor of signal transducer and activator of transcription.

Solubility
Soluble in diochloromethane. Slightly soluble in water.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 57561-39-4
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.228
MDL Number MFCD03425857
InChI Key RFDSJHHLGFFVHD-UHFFFAOYSA-N
Synonym n-boc-n-methyl-aminoethanol, tert-butyl 2-hydroxyethyl methyl carbamate, boc,me-glycinol, tert-butyl n-2-hydroxyethyl-n-methylcarbamate, 2-n-methyl-n-boc-amino ethanol, 2-n-boc-n-methylamino ethanol, n-boc-n-methylethanolamine, 2-hydroxyethyl methylcarbamic acid 1,1-dimethylethyl ester, carbamic acid, n-2-hydroxyethyl-n-methyl-, 1,1-dimethylethyl ester
PubChem CID 545700
IUPAC Name tert-butyl N-(2-hydroxyethyl)-N-methylcarbamate
SMILES CC(C)(C)OC(=O)N(C)CCO

Specifications

Color Colorless to Yellow
Beilstein 3537964
Solubility Information Soluble in diochloromethane. Slightly soluble in water.
Quantity 1 g
Formula Weight 175.23
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-Boc-N-methylethanolamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only