missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-L-serine, 97%

CAS: 101772-29-6 | C9H17NO5 | 219.237 g/mol

$196.73 - $887.89

Chemical Identifiers

CAS 101772-29-6
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00037249
InChI Key NJQGIDVCNBZXLG-LURJTMIESA-N
Synonym boc-n-me-ser-oh, boc-n-methyl-l-serine, boc-n-a-methyl-l-serine, boc-n-, a-methyl-l-serine, s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, n-boc-n-methyl-l-serine, l-serine,n-1,1-dimethylethoxy carbonyl-n-methyl, boc-meser-oh, 2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, boc-l-meser-oh
PubChem CID 7009594
IUPAC Name (2S)-3-hydroxy-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
SMILES CC(C)(C)OC(=O)N(C)C(CO)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH6331403
View Documents Promotion Details
N/A 1 g
Each for $196.73
Only null left
 
AAH6331406
View Documents Promotion Details
N/A 5 g
Each for $887.89
Only null left
 
Description

Description

N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

101772-29-6
219.237
NJQGIDVCNBZXLG-LURJTMIESA-N
7009594
CC(C)(C)OC(=O)N(C)C(CO)C(=O)O
C9H17NO5
MFCD00037249
boc-n-me-ser-oh, boc-n-methyl-l-serine, boc-n-a-methyl-l-serine, boc-n-, a-methyl-l-serine, s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, n-boc-n-methyl-l-serine, l-serine,n-1,1-dimethylethoxy carbonyl-n-methyl, boc-meser-oh, 2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, boc-l-meser-oh
(2S)-3-hydroxy-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid

Specifications

83°C to 87°C
C9H17NO5
8685334
NJQGIDVCNBZXLG-LURJTMIESA-N
(2S)-3-hydroxy-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
7009594
98%
N-Boc-N-methyl-L-serine
101772-29-6
MFCD00037249
boc-n-me-ser-oh, boc-n-methyl-l-serine, boc-n-a-methyl-l-serine, boc-n-, a-methyl-l-serine, s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, n-boc-n-methyl-l-serine, l-serine,n-1,1-dimethylethoxy carbonyl-n-methyl, boc-meser-oh, 2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, boc-l-meser-oh
CC(C)(C)OC(=O)N(C)C(CO)C(=O)O
219.237
219.23
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

RUO – Research Use Only