missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-L-serine, 97%

Catalog No. AAH6331403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6331403 1 g
AAH6331406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6331403 Supplier Thermo Scientific Chemicals Supplier No. H6331403

CAS: 101772-29-6 | C9H17NO5 | 219.237 g/mol

N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-N-methyl-L-serine is used as a precursor in organic synthesis and pharmaceuticals.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 101772-29-6
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00037249
InChI Key NJQGIDVCNBZXLG-LURJTMIESA-N
Synonym boc-n-me-ser-oh, boc-n-methyl-l-serine, boc-n-a-methyl-l-serine, boc-n-, a-methyl-l-serine, s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, n-boc-n-methyl-l-serine, l-serine,n-1,1-dimethylethoxy carbonyl-n-methyl, boc-meser-oh, 2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid, boc-l-meser-oh
PubChem CID 7009594
IUPAC Name (2S)-3-hydroxy-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
SMILES CC(C)(C)OC(=O)N(C)C(CO)C(=O)O

Specifications

Melting Point 83°C to 87°C
Beilstein 8685334
Quantity 1 g
Formula Weight 219.23
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-N-methyl-L-serine

RUO – Research Use Only