missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-L-phenylalanine, 95%

Catalog No. AAH6222103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6222103 1 g
AAH6222106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6222103 Supplier Thermo Scientific Chemicals Supplier No. H6222103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 37553-65-4 | C15H21NO4 | 279.336 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37553-65-4
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.336
MDL Number MFCD00038754
InChI Key AJGJINVEYVTDNH-LBPRGKRZSA-N
Synonym boc-n-me-phe-oh, boc-n-methyl-l-phenylalanine, boc-mephe-oh, boc-l-mephe-oh, phenylalanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, bocmepheoh, ambotzbaa1113, boc me phe oh, pubchem12252
PubChem CID 7010602
IUPAC Name (2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)N(C)C(CC1=CC=CC=C1)C(=O)O

Specifications

Beilstein 2386503
Quantity 1 g
Formula Weight 279.33
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-N-methyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only