CAS RN 37553-65-4
IUPAC Name:
2-{[(tert-butoxy)carbonyl](methyl)amino}-3-phenylpropanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)(methyl)amino]-3-phenylpropanoic acid
Molecular Weight (g/mol):
279.34
Molecular Formula:
C15H21NO4
InChi Key:
AJGJINVEYVTDNH-UHFFFAOYNA-N
SMILES:
CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
Promotions Available
N-Boc-N-methyl-L-phenylalanine, 95%
N-Boc-N-methyl-L-phenylalanine, 95%
CAS: 37553-65-4 Molecular Formula: C15H21NO4 Molecular Weight (g/mol): 279.336 MDL Number: MFCD00038754 InChI Key: AJGJINVEYVTDNH-LBPRGKRZSA-N Synonym: boc-n-me-phe-oh,boc-n-methyl-l-phenylalanine,boc-mephe-oh,boc-l-mephe-oh,phenylalanine, n-1,1-dimethylethoxy carbonyl-n-methyl,2s-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid,bocmepheoh,ambotzbaa1113,boc me phe oh,pubchem12252 PubChem CID: 7010602 IUPAC Name: (2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]-3-phenylpropanoic acid SMILES: CC(C)(C)OC(=O)N(C)C(CC1=CC=CC=C1)C(=O)O