missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-1,3-diaminopropane, 95%, Thermo Scientific Chemicals

$93.56 - $261.26

Chemical Identifiers

CAS 150349-36-3
Molecular Formula C9H20N2O2
Molecular Weight (g/mol) 188.271
MDL Number MFCD01631189
InChI Key PNQYAMWGTGWJDW-UHFFFAOYSA-N
Synonym n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
PubChem CID 2756543
IUPAC Name tert-butyl N-(3-aminopropyl)-N-methylcarbamate
SMILES CC(C)(C)OC(=O)N(C)CCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66932MD
View Documents
Thermo Scientific Chemicals
H66932MD
250 mg
Each for $93.56
Only null left
 
AAH6693203
View Documents
Thermo Scientific Chemicals
H6693203
1 g
Each for $261.26
Only null left
 
Description

Description

It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

150349-36-3
188.271
PNQYAMWGTGWJDW-UHFFFAOYSA-N
2756543
CC(C)(C)OC(=O)N(C)CCCN
C9H20N2O2
MFCD01631189
n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
tert-butyl N-(3-aminopropyl)-N-methylcarbamate

Specifications

150349-36-3
C9H20N2O2
n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
PNQYAMWGTGWJDW-UHFFFAOYSA-N
tert-butyl N-(3-aminopropyl)-N-methylcarbamate
188.271
188.27
Liquid
Yellow
MFCD01631189
Slightly soluble in water.
CC(C)(C)OC(=O)N(C)CCCN
250 mg
2756543
95%
N-Boc-N-methyl-1,3-diaminopropane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only