missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-1,3-diaminopropane, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6693203

 View more versions of this product

Catalog No. AAH6693203


Only null left
Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

N-Boc-N-methyl-1,3-diaminopropane
Yellow
MFCD01631189
Slightly soluble in water.
CC(C)(C)OC(=O)N(C)CCCN
1 g
2756543
95%
150349-36-3
C9H20N2O2
n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
PNQYAMWGTGWJDW-UHFFFAOYSA-N
tert-butyl N-(3-aminopropyl)-N-methylcarbamate
188.271
188.27
Liquid

Chemical Identifiers

150349-36-3
188.271
PNQYAMWGTGWJDW-UHFFFAOYSA-N
2756543
CC(C)(C)OC(=O)N(C)CCCN
C9H20N2O2
MFCD01631189
n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
tert-butyl N-(3-aminopropyl)-N-methylcarbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only