missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-tryptophanol, 95%, Thermo Scientific Chemicals

$58.67 - $58.67

Chemical Identifiers

CAS 82689-19-8
Molecular Formula C16H22N2O3
Molecular Weight (g/mol) 290.36
MDL Number MFCD00235953
InChI Key JEFQUFUAEKORKL-LBPRGKRZSA-N
Synonym n-boc-l-tryptophanol, boc-tryptophanol, boc-l-tryptophanol, boc-trp-ol, s-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, s-3-2-boc-amino-3-hydroxypropyl-indole, boc-d-trp-ol, n-alpha-boc-l-tryptophanol, boc-trypotophanol, boc-l-trp-ol
PubChem CID 7019533
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CO)CC1=CNC2=CC=CC=C12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66571MD
View Documents
Thermo Scientific Chemicals
H66571MD
250 mg
Each for $58.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

82689-19-8
290.36
JEFQUFUAEKORKL-LBPRGKRZSA-N
7019533
CC(C)(C)OC(=O)N[C@H](CO)CC1=CNC2=CC=CC=C12
C16H22N2O3
MFCD00235953
n-boc-l-tryptophanol, boc-tryptophanol, boc-l-tryptophanol, boc-trp-ol, s-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, s-3-2-boc-amino-3-hydroxypropyl-indole, boc-d-trp-ol, n-alpha-boc-l-tryptophanol, boc-trypotophanol, boc-l-trp-ol
tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate

Specifications

118°C to 122°C
C16H22N2O3
n-boc-l-tryptophanol, boc-tryptophanol, boc-l-tryptophanol, boc-trp-ol, s-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, s-3-2-boc-amino-3-hydroxypropyl-indole, boc-d-trp-ol, n-alpha-boc-l-tryptophanol, boc-trypotophanol, boc-l-trp-ol
CC(C)(C)OC(=O)N[C@H](CO)CC1=CNC2=CC=CC=C12
250 mg
7019533
95%
N-Boc-L-tryptophanol
82689-19-8
MFCD00235953
JEFQUFUAEKORKL-LBPRGKRZSA-N
tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
290.36
290.36
Solid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only