missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-threonine methyl ester, 95%

CAS: 79479-07-5 | C10H19NO5 | 233.26 g/mol

$126.93 - $517.07

Chemical Identifiers

CAS 79479-07-5
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.26
MDL Number MFCD00270479
InChI Key MZMWAPNVRMDIPS-RQJHMYQMSA-N
Synonym boc-thr-ome, 2s,3r-methyl 2-tert-butoxycarbonyl amino-3-hydroxybutanoate, boc-l-threonine methyl ester, n-boc-l-threonine methyl ester, n-tert-butoxycarbonyl-l-threonine methyl ester, n-tert-butoxycarbonyl threonine methyl ester, methyl 2s,3r-2-tert-butoxycarbonyl amino-3-hydroxybutanoate, methyl n-tert-butoxycarbonyl-l-threoninate, n-t-butoxycarbonyl-l-threonine methyl ester
PubChem CID 11368286
IUPAC Name methyl (2S,3R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate
SMILES COC(=O)[C@@H](NC(=O)OC(C)(C)C)[C@@H](C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAH6154506
View Documents
Thermo Scientific Chemicals
H6154506
5 g
Each for $126.93
Only null left
 
AAH6154514
View Documents
Thermo Scientific Chemicals
H6154514
25 g
Each for $517.07
Only null left
 
Description

Description

N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Storage temperature 2 - 4°C. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

79479-07-5
233.26
MZMWAPNVRMDIPS-RQJHMYQMSA-N
11368286
COC(=O)[C@@H](NC(=O)OC(C)(C)C)[C@@H](C)O
C10H19NO5
MFCD00270479
boc-thr-ome, 2s,3r-methyl 2-tert-butoxycarbonyl amino-3-hydroxybutanoate, boc-l-threonine methyl ester, n-boc-l-threonine methyl ester, n-tert-butoxycarbonyl-l-threonine methyl ester, n-tert-butoxycarbonyl threonine methyl ester, methyl 2s,3r-2-tert-butoxycarbonyl amino-3-hydroxybutanoate, methyl n-tert-butoxycarbonyl-l-threoninate, n-t-butoxycarbonyl-l-threonine methyl ester
methyl (2S,3R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate

Specifications

79479-07-5
Yellow
1.45
boc-thr-ome, 2s,3r-methyl 2-tert-butoxycarbonyl amino-3-hydroxybutanoate, boc-l-threonine methyl ester, n-boc-l-threonine methyl ester, n-tert-butoxycarbonyl-l-threonine methyl ester, n-tert-butoxycarbonyl threonine methyl ester, methyl 2s,3r-2-tert-butoxycarbonyl amino-3-hydroxybutanoate, methyl n-tert-butoxycarbonyl-l-threoninate, n-t-butoxycarbonyl-l-threonine methyl ester
MZMWAPNVRMDIPS-RQJHMYQMSA-N
methyl (2S,3R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate
233.26
233.26
Viscous Liquid
206°C
C10H19NO5
MFCD00270479
Slightly soluble in water.
COC(=O)[C@@H](NC(=O)OC(C)(C)C)[C@@H](C)O
5 g
11368286
95%
N-Boc-L-threonine methyl ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only