accessibility menu, dialog, popup

UserName

(R)CH3OO(S)HNH3COHOOCH3CH3CH3

CAS RN 79479-07-5

IUPAC Name: methyl (2S,3R)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxybutanoate
Synonyms: methyl (2S,3R)-2-[(tert-butoxycarbonyl)amino]-3-hydroxybutanoate|BOC-THR-OME
Molecular Weight (g/mol): 233.26
Molecular Formula: C10H19NO5
InChi Key: MZMWAPNVRMDIPS-RQJHMYQMSA-N
SMILES: COC(=O)[C@@H](NC(=O)OC(C)(C)C)[C@@H](C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)