missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-threonine methyl ester, 95%

Catalog No. AAH6154506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6154506 5 g
AAH6154514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6154506 Supplier Thermo Scientific Chemicals Supplier No. H6154506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79479-07-5 | C10H19NO5 | 233.26 g/mol

N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Storage temperature 2 - 4°C. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 79479-07-5
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.26
MDL Number MFCD00270479
InChI Key MZMWAPNVRMDIPS-RQJHMYQMSA-N
Synonym boc-thr-ome, 2s,3r-methyl 2-tert-butoxycarbonyl amino-3-hydroxybutanoate, boc-l-threonine methyl ester, n-boc-l-threonine methyl ester, n-tert-butoxycarbonyl-l-threonine methyl ester, n-tert-butoxycarbonyl threonine methyl ester, methyl 2s,3r-2-tert-butoxycarbonyl amino-3-hydroxybutanoate, methyl n-tert-butoxycarbonyl-l-threoninate, n-t-butoxycarbonyl-l-threonine methyl ester
PubChem CID 11368286
IUPAC Name methyl (2S,3R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate
SMILES COC(=O)[C@@H](NC(=O)OC(C)(C)C)[C@@H](C)O

Specifications

Boiling Point 206°C
Color Yellow
Refractive Index 1.45
Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 233.26
Percent Purity 95%
Physical Form Viscous Liquid
Chemical Name or Material N-Boc-L-threonine methyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only