missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-serine, 98% (dry wt.), may cont. up to 10% water

Catalog No. p-7023137
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1622406 5 g
AAA1622414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1622406 Supplier Thermo Scientific Chemicals Supplier No. A1622406

CAS: 3262-72-4 | C8H15NO5 | 205.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3262-72-4
Molecular Formula C8H15NO5
Molecular Weight (g/mol) 205.21
MDL Number MFCD00037243
InChI Key FHOAKXBXYSJBGX-MLHKIVSYNA-N
Synonym boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser
PubChem CID 98766
SMILES CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O

Specifications

Melting Point 90°C to 91°C (decomposition)
Quantity 5 g
Beilstein 2212252
Optical Rotation −3.5° (c=2 in Acetic Acid)
IUPAC Name (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Formula Weight 205.21
Percent Purity 98%
Assay (dry wt.), may cont. up to 10% water
Chemical Name or Material N-Boc-L-serine
EINECSNumber 221-867-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only