missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-prolinol, 98+%

CAS: 69610-40-8 | C10H19NO3 | 201.266 g/mol

Supplier:  Thermo Scientific Chemicals L0988506

 View more versions of this product

Catalog No. AAL0988506


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Boc-L-prolinol
59°C to 62°C
MFCD00066232
3542667
BFFLLBPMZCIGRM-QMMMGPOBSA-N
CC(C)(C)OC(=O)N1CCCC1CO
201.266
201.27
69610-40-8
C10H19NO3
5 g
n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
−54° (c=5 in Methanol)
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
643448
≥98%

Chemical Identifiers

69610-40-8
201.266
BFFLLBPMZCIGRM-QMMMGPOBSA-N
643448
CC(C)(C)OC(=O)N1CCCC1CO
C10H19NO3
MFCD00066232
n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only