missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-prolinol, 98+%

Catalog No. p-7045230
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0988503 1 g
AAL0988506 5 g
AAL0988514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0988503 Supplier Thermo Scientific Chemicals Supplier No. L0988503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69610-40-8 | C10H19NO3 | 201.266 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69610-40-8
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD00066232
InChI Key BFFLLBPMZCIGRM-QMMMGPOBSA-N
Synonym n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
PubChem CID 643448
IUPAC Name tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CO

Specifications

Melting Point 59°C to 62°C
Quantity 1 g
Beilstein 3542667
Optical Rotation −54° (c=5 in Methanol)
Formula Weight 201.27
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-prolinol
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only