missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-phenylalaninol, 99%

CAS: 66605-57-0 | C14H21NO3 | 251.33 g/mol

Supplier:  Thermo Scientific Chemicals B2501906

Quantity 5 g

 View more versions of this product

Catalog No. AAB2501906


Only null left

Description

Description

N-Boc-L-phenylalaninol is employed in the synthesis of biochemically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-phenylalaninol is employed in the synthesis of biochemically active compounds.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications

Chemical Identifiers

66605-57-0
251.33
LDKDMDVMMCXTMO-LBPRGKRZSA-N
2733675
CC(C)(C)OC(=O)N[C@H](CO)CC1=CC=CC=C1
C14H21NO3
MFCD00076976
n-boc-l-phenylalaninol, boc-l-phenylalaninol, boc-phenylalaninol, s-2-boc-amino-3-phenyl-1-propanol, n-tert-butoxycarbonyl-l-phenylalaninol, tert-butyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate, s-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate, s-n-tert-butoxycarbonyl-phenylalaninol, s---2-tert-butoxycarbonylamino-3-phenyl-1-propanol
tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate

Specifications

95°C to 98°C
66605-57-0
MFCD00076976
n-boc-l-phenylalaninol, boc-l-phenylalaninol, boc-phenylalaninol, s-2-boc-amino-3-phenyl-1-propanol, n-tert-butoxycarbonyl-l-phenylalaninol, tert-butyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate, s-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate, s-n-tert-butoxycarbonyl-phenylalaninol, s---2-tert-butoxycarbonylamino-3-phenyl-1-propanol
LDKDMDVMMCXTMO-LBPRGKRZSA-N
tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
5 g
−27.5° ± 2.5° (c=1 in Methanol)
99%
N-Boc-L-phenylalaninol
C14H21NO3
3652004
Insoluble in water.
CC(C)(C)OC(=O)N[C@H](CO)CC1=CC=CC=C1
251.33
2733675
251.33
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions