missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-BOC-L-Phenylalaninal, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 376742500
| Quantity | 250mg |
|---|---|
| Packaging | Glass bottle |
Description
Chemical Identifiers
| 72155-45-4 | |
| 249.31 | |
| ZJTYRNPLVNMVPQ-LBPRGKRZSA-N | |
| 6998013 | |
| CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C=O |
| C14H19NO3 | |
| MFCD00143854 | |
| n-boc-l-phenylalaninal, boc-phe-cho, boc-l-phenylalaninal, n-boc-phenylalaninal, boc-phenylalaninal, s-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate, chembl94327, s---2-tert-butoxycarbonylamino-3-phenylpropanal, s-tert-butyl 1-oxo-3-phenylpropan-2-ylcarbamate, tert-butyl n-2s-1-oxo-3-phenylpropan-2-yl carbamate | |
| tert-butyl N-[(2S)-1-oxo-3-phenylpropan-2-yl]carbamate |
Specifications
| N-BOC-L-Phenylalaninal | |
| 96.0 | |
| Authentic | |
| Glass bottle | |
| 250mg | |
| − 45.00 | |
| ZJTYRNPLVNMVPQ-LBPRGKRZSA-N | |
| tert-butyl N-[(2S)-1-oxo-3-phenylpropan-2-yl]carbamate | |
| 6998013 | |
| 97% |
| 72155-45-4 | |
| 100.0 | |
| 96% min. (GC) | |
| C14H19NO3 | |
| MFCD00143854 | |
| n-boc-l-phenylalaninal, boc-phe-cho, boc-l-phenylalaninal, n-boc-phenylalaninal, boc-phenylalaninal, s-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate, chembl94327, s---2-tert-butoxycarbonylamino-3-phenylpropanal, s-tert-butyl 1-oxo-3-phenylpropan-2-ylcarbamate, tert-butyl n-2s-1-oxo-3-phenylpropan-2-yl carbamate | |
| CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C=O | |
| 249.31 | |
| 249.31 |