CAS RN 72155-45-4
CAS RN 72155-45-4
IUPAC Name:
tert-butyl N-[(2S)-1-oxo-3-phenylpropan-2-yl]carbamate
Synonyms:
tert-butyl N-[(2S)-1-oxo-3-phenylpropan-2-yl]carbamate|N-BOC-L-PHENYLALANINAL
Molecular Weight (g/mol):
249.31
Molecular Formula:
C14H19NO3
InChi Key:
ZJTYRNPLVNMVPQ-LBPRGKRZSA-N
SMILES:
CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C=O
1
–
1
of
1
results
1
N-BOC-L-Phenylalaninal, 97%, Thermo Scientific™
CAS: 72155-45-4 Molecular Formula: C14H19NO3 Molecular Weight (g/mol): 249.31 MDL Number: MFCD00143854 InChI Key: ZJTYRNPLVNMVPQ-LBPRGKRZSA-N Synonym: n-boc-l-phenylalaninal,boc-phe-cho,boc-l-phenylalaninal,n-boc-phenylalaninal,boc-phenylalaninal,s-tert-butyl 1-oxo-3-phenylpropan-2-yl carbamate,chembl94327,s---2-tert-butoxycarbonylamino-3-phenylpropanal,s-tert-butyl 1-oxo-3-phenylpropan-2-ylcarbamate,tert-butyl n-2s-1-oxo-3-phenylpropan-2-yl carbamate PubChem CID: 6998013 IUPAC Name: tert-butyl N-[(2S)-1-oxo-3-phenylpropan-2-yl]carbamate SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C=O