missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-leucinol, 97%

Catalog No. AAL1935206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1935206 5 g
AAL1935203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1935206 Supplier Thermo Scientific Chemicals Supplier No. L1935206

CAS: 82010-31-9 | C11H23NO3 | 217.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82010-31-9
Molecular Formula C11H23NO3
Molecular Weight (g/mol) 217.31
MDL Number MFCD00076950
InChI Key LQTMEOSBXTVYRM-VIFPVBQESA-N
Synonym boc-l-leucinol, n-boc-l-leucinol, boc-leucinol, boc-leu-ol, tert-butyl n-2s-1-hydroxy-4-methylpentan-2-yl carbamate, n-boc l-leucinol, pubchem12195, l-leucinol, n-boc protected, n-t-butoxycarbonyl-l-leucinol
PubChem CID 7018766
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-4-methylpentan-2-yl]carbamate
SMILES CC(C)C[C@@H](CO)NC(=O)OC(C)(C)C

Specifications

Boiling Point 212°C to 214°C
Refractive Index 1.45
Beilstein 4178460
Density 0.98
Quantity 5 g
Flash Point >110°C (230°F)
Optical Rotation −27.5° (c=2 in Methanol)
Formula Weight 217.31
Percent Purity 97%
Chemical Name or Material N-Boc-L-leucinol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only