missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-isoleucinol, 95%, Thermo Scientific Chemicals

$473.53

Chemical Identifiers

CAS 106946-74-1
Molecular Formula C11H23NO3
Molecular Weight (g/mol) 217.31
MDL Number MFCD00235929
InChI Key BPLDQMXXYMKQPW-DTWKUNHWSA-N
Synonym n-boc-l-isoleucinol, boc-isoleucinol, n-boc-l-isolucinole, n-boc-2s,3s---2-amino-3-methyl-1-pentanol, boc-l-isoleucinol, tert-butyl 2s,3s-1-hydroxy-3-methylpentan-2-yl carbamate, boc-ile-ol, ambotzbal1038, pubchem15801
PubChem CID 14237729
IUPAC Name tert-butyl N-[(2S,3S)-1-hydroxy-3-methylpentan-2-yl]carbamate
SMILES CC[C@H](C)[C@@H](CO)NC(=O)OC(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAL1935106
View Documents Promotion Details
N/A 5 g
Each for $473.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106946-74-1
217.31
BPLDQMXXYMKQPW-DTWKUNHWSA-N
14237729
CC[C@H](C)[C@@H](CO)NC(=O)OC(C)(C)C
C11H23NO3
MFCD00235929
n-boc-l-isoleucinol, boc-isoleucinol, n-boc-l-isolucinole, n-boc-2s,3s---2-amino-3-methyl-1-pentanol, boc-l-isoleucinol, tert-butyl 2s,3s-1-hydroxy-3-methylpentan-2-yl carbamate, boc-ile-ol, ambotzbal1038, pubchem15801
tert-butyl N-[(2S,3S)-1-hydroxy-3-methylpentan-2-yl]carbamate

Specifications

106946-74-1
C11H23NO3
MFCD00235929
n-boc-l-isoleucinol, boc-isoleucinol, n-boc-l-isolucinole, n-boc-2s,3s---2-amino-3-methyl-1-pentanol, boc-l-isoleucinol, tert-butyl 2s,3s-1-hydroxy-3-methylpentan-2-yl carbamate, boc-ile-ol, ambotzbal1038, pubchem15801
CC[C@H](C)[C@@H](CO)NC(=O)OC(C)(C)C
217.31
217.31
N-Boc-L-isoleucinol
121°C to 126°C (0.6mmHg)
1.453
3604099
BPLDQMXXYMKQPW-DTWKUNHWSA-N
tert-butyl N-[(2S,3S)-1-hydroxy-3-methylpentan-2-yl]carbamate
14237729
95%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 624-790-7

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only