missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-isoleucinol, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1935106

Quantity 5 g

 View more versions of this product

Catalog No. AAL1935106


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106946-74-1
217.31
BPLDQMXXYMKQPW-DTWKUNHWSA-N
14237729
CC[C@H](C)[C@@H](CO)NC(=O)OC(C)(C)C
C11H23NO3
MFCD00235929
n-boc-l-isoleucinol, boc-isoleucinol, n-boc-l-isolucinole, n-boc-2s,3s---2-amino-3-methyl-1-pentanol, boc-l-isoleucinol, tert-butyl 2s,3s-1-hydroxy-3-methylpentan-2-yl carbamate, boc-ile-ol, ambotzbal1038, pubchem15801
tert-butyl N-[(2S,3S)-1-hydroxy-3-methylpentan-2-yl]carbamate

Specifications

N-Boc-L-isoleucinol
121°C to 126°C (0.6mmHg)
1.453
3604099
BPLDQMXXYMKQPW-DTWKUNHWSA-N
tert-butyl N-[(2S,3S)-1-hydroxy-3-methylpentan-2-yl]carbamate
5 g
217.31
106946-74-1
C11H23NO3
MFCD00235929
n-boc-l-isoleucinol, boc-isoleucinol, n-boc-l-isolucinole, n-boc-2s,3s---2-amino-3-methyl-1-pentanol, boc-l-isoleucinol, tert-butyl 2s,3s-1-hydroxy-3-methylpentan-2-yl carbamate, boc-ile-ol, ambotzbal1038, pubchem15801
CC[C@H](C)[C@@H](CO)NC(=O)OC(C)(C)C
217.31
14237729
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 624-790-7

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions