missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone, Thermo Scientific Chemicals

Catalog No. AAJ65812LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65812LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65812LB0 Supplier Thermo Scientific Chemicals Supplier No. J65812LB0
Only null left

 

N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone is used as a novel Boc-D(OMe)-FMK inhibitor synthesized using proprietary technology and irreversibly binds to activated caspases to block apoptosis. It is useful for in vitro and in vivo applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone is used as a novel Boc-D(OMe)-FMK inhibitor synthesized using proprietary technology and irreversibly binds to activated caspases to block apoptosis. It is useful for in vitro

Chemical Identifiers

CAS 187389-53-3
Molecular Formula C11H18FNO5
Molecular Weight (g/mol) 263.265
MDL Number MFCD03453073
InChI Key MXOOUCRHWJYCAL-ZETCQYMHSA-N
Synonym boc-d-fmk, caspase inhibitor iii, boc-asp ome-fluoromethyl ketone, boc-asp ome-fmk, boc-d ome-fmk, methyl 3-tert-butoxycarbonyl amino-5-fluoro-4-oxopentanoate, caspase3-inhibitor boc-d-fmk, boc-asp ome-fluoromethylketone, caspase inhibitor iii and boc-d-fmk, 3-tert-butoxy carbonyl amino-5-fluoro-4-oxopentanoic acid methyl ester
PubChem CID 9881695
IUPAC Name methyl (3S)-5-fluoro-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxopentanoate
SMILES CC(C)(C)OC(=O)NC(CC(=O)OC)C(=O)CF

Specifications

Color White
Solubility Information Soluble in dry DMSO or DMF at approximately 20mM; Store stock solutions at-20°C.
Quantity 5 mg
Formula Weight 263.26
Physical Form Powder
Chemical Name or Material N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only