accessibility menu, dialog, popup

UserName

CAS RN 187389-53-3

CH3CH3CH3OOHNOOH3COF

CAS RN 187389-53-3

IUPAC Name: methyl 3-{[(tert-butoxy)carbonyl]amino}-5-fluoro-4-oxopentanoate
Synonyms: methyl 3-[(tert-butoxycarbonyl)amino]-5-fluoro-4-oxopentanoate
Molecular Weight (g/mol): 263.27
Molecular Formula: C11H18FNO5
InChi Key: MXOOUCRHWJYCAL-UHFFFAOYNA-N
SMILES: COC(=O)CC(NC(=O)OC(C)(C)C)C(=O)CF

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)