missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-aspartic acid 1-tert-butyl ester, 95%
CAS: 34582-32-6 | C13H23NO6 | 289.328 g/mol
$104.67 - $397.19
Chemical Identifiers
| CAS | 34582-32-6 |
|---|---|
| Molecular Formula | C13H23NO6 |
| Molecular Weight (g/mol) | 289.328 |
| MDL Number | MFCD00038272 |
| InChI Key | RAUQRYTYJIYLTF-QMMMGPOBSA-N |
| Synonym | boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu |
| PubChem CID | 7010517 |
| IUPAC Name | (3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
| SMILES | CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6242503
|
Thermo Scientific Chemicals
H6242503 |
1 g |
Each for $104.67
|
|
|||||
|
AAH6242506
|
Thermo Scientific Chemicals
H6242506 |
5 g |
Each for $397.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 34582-32-6 | |
| 289.328 | |
| RAUQRYTYJIYLTF-QMMMGPOBSA-N | |
| 7010517 | |
| CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C |
| C13H23NO6 | |
| MFCD00038272 | |
| boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu | |
| (3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
Specifications
| 34582-32-6 | |
| C13H23NO6 | |
| 4191701 | |
| RAUQRYTYJIYLTF-QMMMGPOBSA-N | |
| (3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid | |
| 289.328 | |
| Odorless | |
| 95% | |
| N-Boc-L-aspartic acid 1-tert-butyl ester |
| White | |
| MFCD00038272 | |
| boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu | |
| CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C | |
| 1 g | |
| 7010517 | |
| 289.32 | |
| Powder |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only