missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-aspartic acid 1-tert-butyl ester, 95%

CAS: 34582-32-6 | C13H23NO6 | 289.328 g/mol

Supplier:  Thermo Scientific Chemicals H6242503

 View more versions of this product

Catalog No. AAH6242503


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Boc-L-aspartic acid 1-tert-butyl ester
White
MFCD00038272
boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu
CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C
1 g
7010517
289.32
Powder
34582-32-6
C13H23NO6
4191701
RAUQRYTYJIYLTF-QMMMGPOBSA-N
(3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
289.328
Odorless
95%

Chemical Identifiers

34582-32-6
289.328
RAUQRYTYJIYLTF-QMMMGPOBSA-N
7010517
CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OC(C)(C)C
C13H23NO6
MFCD00038272
boc-asp-otbu, boc-l-aspartic acid 1-tert-butyl ester, s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-tert-boc-l-aspartic acid tert-butyl ester, 3s-4-tert-butoxy-3-tert-butoxycarbonylamino-4-oxo-butanoic acid, 3s-4-tert-butoxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp-otbu, boc-asp-o-t-butyl, pubchem12140, boc-aspartic acid-otbu
(3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only