missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycinamide, 95%

Catalog No. AAH6205306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6205306 5 g
AAH6205303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6205306 Supplier Thermo Scientific Chemicals Supplier No. H6205306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35150-09-5 | C7H14N2O3 | 174.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35150-09-5
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.2
MDL Number MFCD08275106
InChI Key RHONTQZNLFIDCQ-UHFFFAOYSA-N
Synonym boc-glycinamide, tert-butyl 2-amino-2-oxoethyl carbamate, boc-gly-nh2, tert-butyl n-carbamoylmethyl carbamate, n-tert-butoxycarbonyl-l-glycinamide, carbamic acid, 2-amino-2-oxoethyl-, 1,1-dimethylethyl ester, carbamoylmethyl-carbamic acid tert-butyl ester, boc-glycine amide, t-butyl 2-amino-2-oxoethyl carbamate
PubChem CID 10910064
IUPAC Name tert-butyl N-(2-amino-2-oxoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC(=O)N

Specifications

Color White
Linear Formula (CH3)3COCONHCH2CONH2
Quantity 5 g
Formula Weight 174.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-glycinamide
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only