CAS RN 35150-09-5
IUPAC Name:
tert-butyl N-(carbamoylmethyl)carbamate
Synonyms:
tert-butyl N-(carbamoylmethyl)carbamate
Molecular Weight (g/mol):
174.2
Molecular Formula:
C7H14N2O3
InChi Key:
RHONTQZNLFIDCQ-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NCC(N)=O
1
–
1
of
1
results
1
Promotions Available
N-Boc-glycinamide, 95%
N-Boc-glycinamide, 95%
CAS: 35150-09-5 Molecular Formula: C7H14N2O3 Molecular Weight (g/mol): 174.2 MDL Number: MFCD08275106 InChI Key: RHONTQZNLFIDCQ-UHFFFAOYSA-N Synonym: boc-glycinamide,tert-butyl 2-amino-2-oxoethyl carbamate,boc-gly-nh2,tert-butyl n-carbamoylmethyl carbamate,n-tert-butoxycarbonyl-l-glycinamide,carbamic acid, 2-amino-2-oxoethyl-, 1,1-dimethylethyl ester,carbamoylmethyl-carbamic acid tert-butyl ester,boc-glycine amide,t-butyl 2-amino-2-oxoethyl carbamate PubChem CID: 10910064 IUPAC Name: tert-butyl N-(2-amino-2-oxoethyl)carbamate SMILES: CC(C)(C)OC(=O)NCC(=O)N