missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-DL-pipecolinic acid, 98%

CAS: 98303-20-9 | C11H19NO4 | 229.276 g/mol

$40.25 - $88.62

Chemical Identifiers

CAS 98303-20-9
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD01862877
InChI Key JQAOHGMPAAWWQO-UHFFFAOYSA-N
Synonym n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid
PubChem CID 581831
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1752803
View Documents
Thermo Scientific Chemicals
L1752803
1 g
Each for $40.25
Only null left
 
AAL1752806
View Documents
Thermo Scientific Chemicals
L1752806
5 g
Each for $88.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98303-20-9
229.276
JQAOHGMPAAWWQO-UHFFFAOYSA-N
581831
CC(C)(C)OC(=O)N1CCCCC1C(=O)O
C11H19NO4
MFCD01862877
n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid
1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid

Specifications

98303-20-9
C11H19NO4
1 g
n-boc-2-piperidinecarboxylic acid, 1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 1-boc-piperidine-2-carboxylic acid, n-boc-dl-pipecolinic acid, n-boc-piperidine-2-carboxylic acid, boc-dl-pip-oh, n-boc-pipecolic acid, 1-n-boc-2-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-2-carboxylic acid, 1-tert-butoxycarbonyl-2-piperidinecarboxylic acid
CC(C)(C)OC(=O)N1CCCCC1C(=O)O
229.276
229.28
N-Boc-DL-pipecolinic acid
129°C to 131°C
MFCD01862877
480022
JQAOHGMPAAWWQO-UHFFFAOYSA-N
1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
581831
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only