accessibility menu, dialog, popup

UserName

CAS RN 98303-20-9

IUPAC Name: (2S)-1-[(tert-butoxy)carbonyl]piperidine-2-carboxylate
Synonyms: (2S)-1-(tert-butoxycarbonyl)piperidine-2-carboxylate
Molecular Weight (g/mol): 228.27
Molecular Formula: C11H18NO4
InChi Key: JQAOHGMPAAWWQO-QMMMGPOBSA-M
SMILES: CC(C)(C)OC(=O)N1CCCC[C@H]1C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results