missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-valine, 98+%

Catalog No. AAL0919303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0919303 1 g
AAL0919306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0919303 Supplier Thermo Scientific Chemicals Supplier No. L0919303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22838-58-0 | C10H19NO4 | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22838-58-0
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00038282
InChI Key SZXBQTSZISFIAO-SSDOTTSWSA-N
Synonym boc-d-valine, boc-d-val-oh, n-boc-d-valine, n-tert-butoxycarbonyl-d-valine, tert-butoxycarbonyl-d-valine, r-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, d-valine, n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-valin, r-2-tert-butoxycarbonylamino-3-methylbutanoic acid, n-alpha-t-butyloxycarbonyl-d-valine
PubChem CID 637605
IUPAC Name (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 76°C to 80°C
Quantity 1 g
Beilstein 2050408
Optical Rotation +6.3° (c=1 in Acetic Acid)
Formula Weight 217.27
Percent Purity ≥98%
Chemical Name or Material N-Boc-D-valine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only