missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iodide, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7164996
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608836 125 g Amber Glass Bottle
18608838 500 g Amber Glass Bottle
18608693 2.5 kg Amber Glass Bottle
18608837 12 kg Amber Glass Bottle
18608839 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608836 Supplier Spectrum Chemical Mfg Cor Supplier No. A1205125GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
IUPAC Name ammonium iodide
SMILES [NH4+].[I-]

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 125 g
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle
Residue after Ignition 0.0005
Chemical Name or Material Ammonium Iodide

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.