missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-serine benzyl ester, 95%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 141527-78-8
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00080265
InChI Key BQADRZHPZVQGCW-GFCCVEGCSA-N
Synonym boc-d-ser-obzl, boc-d-ser-o-bzl, boc-d-serine benzyl ester, r-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, benzyl n-tert-butoxycarbonyl-d-serinate, benzyl 2r-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-d-serine benzylester, boc-d-ser-o-bzl hplc
PubChem CID 12953536
IUPAC Name benzyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH6651406
View Documents Promotion Details
N/A 5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141527-78-8
295.335
BQADRZHPZVQGCW-GFCCVEGCSA-N
12953536
CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1
C15H21NO5
MFCD00080265
boc-d-ser-obzl, boc-d-ser-o-bzl, boc-d-serine benzyl ester, r-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, benzyl n-tert-butoxycarbonyl-d-serinate, benzyl 2r-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-d-serine benzylester, boc-d-ser-o-bzl hplc
benzyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate

Specifications

66°C to 70°C
White
MFCD00080265
BQADRZHPZVQGCW-GFCCVEGCSA-N
benzyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
12953536
95%
N-Boc-D-serine benzyl ester
141527-78-8
C15H21NO5
boc-d-ser-obzl, boc-d-ser-o-bzl, boc-d-serine benzyl ester, r-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, benzyl n-tert-butoxycarbonyl-d-serinate, benzyl 2r-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-d-serine benzylester, boc-d-ser-o-bzl hplc
CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1
295.335
295.34
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only