missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-serine benzyl ester, 95%, Thermo Scientific Chemicals

Catalog No. AAH6651406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6651406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6651406 Supplier Thermo Scientific Chemicals Supplier No. H6651406
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141527-78-8
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00080265
InChI Key BQADRZHPZVQGCW-GFCCVEGCSA-N
Synonym boc-d-ser-obzl, boc-d-ser-o-bzl, boc-d-serine benzyl ester, r-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, benzyl n-tert-butoxycarbonyl-d-serinate, benzyl 2r-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, boc-d-serine benzylester, boc-d-ser-o-bzl hplc
PubChem CID 12953536
IUPAC Name benzyl (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 66°C to 70°C
Color White
Quantity 5 g
Formula Weight 295.34
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-D-serine benzyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only