missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-glutamic acid 1-tert-butyl ester, 95%

Catalog No. AAH6232003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6232003 1 g
AAH6232006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6232003 Supplier Thermo Scientific Chemicals Supplier No. H6232003
Only null left

CAS: 73872-71-6 | C14H25NO6 | 303.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73872-71-6
Molecular Formula C14H25NO6
Molecular Weight (g/mol) 303.36
MDL Number MFCD00076926
InChI Key YMOYURYWGUWMFM-UHFFFAOYNA-N
Synonym boc-d-glu-otbu, r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, 4r-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-o-t-bu, n-boc-d-glutamic acid 1-tert-butyl ester, d-glutamicacid,n-1,1-dimethylethoxy carbonyl-,1-1,1-dimethylethyl ester
PubChem CID 7018746
SMILES CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C

Specifications

Color Brown to White
IUPAC Name 5-(tert-butoxy)-4-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Quantity 1 g
Formula Weight 303.35
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-D-glutamic acid 1-tert-butyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only