accessibility menu, dialog, popup

UserName

CAS RN 73872-71-6

CH3CH3H3COOOHONHOOH3CCH3H3C

CAS RN 73872-71-6

IUPAC Name: 5-(tert-butoxy)-4-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Synonyms: 5-(tert-butoxy)-4-[(tert-butoxycarbonyl)amino]-5-oxopentanoic acid
Molecular Weight (g/mol): 303.36
Molecular Formula: C14H25NO6
InChi Key: YMOYURYWGUWMFM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)