missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-aspartic acid 1-methyl ester, 97%

CAS: 137130-65-5 | C10H17NO6 | 247.25 g/mol

$164.00 - $549.01

Chemical Identifiers

CAS 137130-65-5
Molecular Formula C10H17NO6
Molecular Weight (g/mol) 247.25
MDL Number MFCD03094776
InChI Key IWFIVTBTZUCTQH-UHFFFAOYNA-N
Synonym boc-d-asp-ome, 3r-3-tert-butoxycarbonyl amino-4-methoxy-4-oxobutanoic acid
PubChem CID 10538431
IUPAC Name 3-{[(tert-butoxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid
SMILES COC(=O)C(CC(O)=O)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6657306
View Documents
Thermo Scientific Chemicals
H6657306
5 g
Each for $164.00
Only null left
 
AAH6657314
View Documents
Thermo Scientific Chemicals
H6657314
25 g
Each for $549.01
Only null left
 
Description

Description

N-Boc-D-aspartic acid 1-methyl ester is widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-D-aspartic acid 1-methyl ester is widely used in pharmaceuticals and foods.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications

Chemical Identifiers

137130-65-5
247.25
IWFIVTBTZUCTQH-UHFFFAOYNA-N
10538431
COC(=O)C(CC(O)=O)NC(=O)OC(C)(C)C
C10H17NO6
MFCD03094776
boc-d-asp-ome, 3r-3-tert-butoxycarbonyl amino-4-methoxy-4-oxobutanoic acid
3-{[(tert-butoxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid

Specifications

137130-65-5
C10H17NO6
boc-d-asp-ome, 3r-3-tert-butoxycarbonyl amino-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC(O)=O)NC(=O)OC(C)(C)C
5 g
10538431
97%
N-Boc-D-aspartic acid 1-methyl ester
White
MFCD03094776
IWFIVTBTZUCTQH-UHFFFAOYNA-N
3-{[(tert-butoxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid
247.25
247.25
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only