Learn More
N-Boc-D-aspartic acid 1-methyl ester, 97%
CAS: 137130-65-5 | C10H17NO6 | 247.25 g/mol
Supplier: Thermo Scientific Chemicals H6657306
Description
N-Boc-D-aspartic acid 1-methyl ester is widely used in pharmaceuticals and foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-D-aspartic acid 1-methyl ester is widely used in pharmaceuticals and foods.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
| N-Boc-D-aspartic acid 1-methyl ester | |
| White | |
| MFCD03094776 | |
| IWFIVTBTZUCTQH-UHFFFAOYNA-N | |
| 3-{[(tert-butoxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid | |
| 247.25 | |
| 247.25 | |
| Powder |
| 137130-65-5 | |
| C10H17NO6 | |
| boc-d-asp-ome, 3r-3-tert-butoxycarbonyl amino-4-methoxy-4-oxobutanoic acid | |
| COC(=O)C(CC(O)=O)NC(=O)OC(C)(C)C | |
| 5 g | |
| 10538431 | |
| 97% |
Chemical Identifiers
| 137130-65-5 | |
| 247.25 | |
| IWFIVTBTZUCTQH-UHFFFAOYNA-N | |
| 10538431 | |
| COC(=O)C(CC(O)=O)NC(=O)OC(C)(C)C |
| C10H17NO6 | |
| MFCD03094776 | |
| boc-d-asp-ome, 3r-3-tert-butoxycarbonyl amino-4-methoxy-4-oxobutanoic acid | |
| 3-{[(tert-butoxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only