Learn More
N-Boc-cis-4-hydroxy-D-proline methyl ester, 95%
CAS: 114676-69-6 | C11H19NO5 | 245.275 g/mol
$126.22 - $489.21
Chemical Identifiers
| CAS | 114676-69-6 |
|---|---|
| Molecular Formula | C11H19NO5 |
| Molecular Weight (g/mol) | 245.275 |
| MDL Number | MFCD00797548 |
| InChI Key | MZMNEDXVUJLQAF-HTQZYQBOSA-N |
| Synonym | 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-d-prolinate, n-boc-cis-4-hydroxy-d-proline methyl ester, 1-tert-butyl 2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl 2r,4r-1-boc-4-hydroxypyrrolidine-2-carboxylate, 2r,4r-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine, o1-tert-butyl o2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, pubchem14128, boc-cis-4-d-hyp-ome |
| PubChem CID | 6951202 |
| IUPAC Name | 1-O-tert-butyl 2-O-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate |
| SMILES | CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6313603
|
Thermo Scientific Chemicals
H6313603 |
1 g |
Each for $126.22
|
|
|||||
|
AAH6313606
|
Thermo Scientific Chemicals
H6313606 |
5 g |
Each for $489.21
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 114676-69-6 | |
| 245.275 | |
| MZMNEDXVUJLQAF-HTQZYQBOSA-N | |
| 6951202 | |
| CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O |
| C11H19NO5 | |
| MFCD00797548 | |
| 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-d-prolinate, n-boc-cis-4-hydroxy-d-proline methyl ester, 1-tert-butyl 2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl 2r,4r-1-boc-4-hydroxypyrrolidine-2-carboxylate, 2r,4r-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine, o1-tert-butyl o2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, pubchem14128, boc-cis-4-d-hyp-ome | |
| 1-O-tert-butyl 2-O-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate |
Specifications
| 114676-69-6 | |
| C11H19NO5 | |
| 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-d-prolinate, n-boc-cis-4-hydroxy-d-proline methyl ester, 1-tert-butyl 2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl 2r,4r-1-boc-4-hydroxypyrrolidine-2-carboxylate, 2r,4r-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine, o1-tert-butyl o2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, pubchem14128, boc-cis-4-d-hyp-ome | |
| CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O | |
| 1 g | |
| 6951202 | |
| 95% | |
| N-Boc-cis-4-hydroxy-D-proline methyl ester |
| White | |
| MFCD00797548 | |
| MZMNEDXVUJLQAF-HTQZYQBOSA-N | |
| 1-O-tert-butyl 2-O-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate | |
| 245.275 | |
| 245.27 | |
| Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only