accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOOH3COH

CAS RN 114676-69-6

IUPAC Name: 1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 245.28
Molecular Formula: C11H19NO5
InChi Key: MZMNEDXVUJLQAF-UHFFFAOYNA-N
SMILES: COC(=O)C1CC(O)CN1C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)