missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-cis-4-hydroxy-D-proline methyl ester, 95%

Catalog No. AAH6313606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6313606 5 g
AAH6313603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6313606 Supplier Thermo Scientific Chemicals Supplier No. H6313606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 114676-69-6 | C11H19NO5 | 245.275 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114676-69-6
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.275
MDL Number MFCD00797548
InChI Key MZMNEDXVUJLQAF-HTQZYQBOSA-N
Synonym 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-d-prolinate, n-boc-cis-4-hydroxy-d-proline methyl ester, 1-tert-butyl 2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl 2r,4r-1-boc-4-hydroxypyrrolidine-2-carboxylate, 2r,4r-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 2r,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine, o1-tert-butyl o2-methyl 2r,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, pubchem14128, boc-cis-4-d-hyp-ome
PubChem CID 6951202
IUPAC Name 1-O-tert-butyl 2-O-methyl (2R,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O

Specifications

Color White
Quantity 5 g
Formula Weight 245.27
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-cis-4-hydroxy-D-proline methyl ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only